missing translation for 'onlineSavingsMsg'
Learn More

Piperacillin sodium salt, 835-1007^mg/mg

Catalog No. AAJ6641909
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6641909 10 g
AAJ6641903 1 g
AAJ6641906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6641909 Supplier Thermo Scientific Chemicals Supplier No. J6641909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria. Piperacillin sodium salt is an antibiotic. It is used in pharmacokinetic studies to optimize antimicrobial therapy and in hypersensitivity reactions. | CAS: 59703-84-3 | C23H26N5NaO7S | 539.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in ethanol, water and methanol.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Store in cool place. Incompatible with acids, heavy metal salts and oxidizing agents.

Chemical Identifiers

CAS 59703-84-3
Molecular Formula C23H26N5NaO7S
Molecular Weight (g/mol) 539.54
MDL Number MFCD00917471
InChI Key WCMIIGXFCMNQDS-QMJSIVKPSA-M
PubChem CID 23666879
ChEBI CHEBI:8233
IUPAC Name sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O

Specifications

Type Piperacillin Sodium Salt
Physical Form Powder
Melting Point 183°C to 185°C
Packaging Glass Bottle
Quantity 10 g
Boiling Point 793°C
Beilstein 5373920
Solubility Information Soluble in ethanol,water and methanol.
Formula Weight 539.54
Chemical Name or Material Piperacillin sodium salt
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 261-868-6
RTECSNumber XI0180000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only