missing translation for 'onlineSavingsMsg'
Learn More

Piperacillin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015111
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015111 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015111 Supplier Merck Emd Millipore Supplier No. PHR18051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 66258-76-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H27N5O7 S · H2O
Synonym Piperacillin monohydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 535.57
Grade Certified Reference Material