missing translation for 'onlineSavingsMsg'
Learn More

(-)-α-Pinene, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111020201
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020201 1 mL
111018568 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020201 Supplier Merck Emd Millipore Supplier No. 805991ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(−)-α-Pinene is a volatile mono-terpenoid compound, that is found in the resins of pine trees and other conifers. It contains an endocyclic double bond in its structure.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 7785-26-4
Density 0.858 g/mL (at 20°C)
Flash Point 33°C
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00064145
Boiling Point 156°C to 158°C
Synonym (1 S,5 S)-2,6,6-Trimethylbicyclo[3.1.1 ]hept-2-ene; (1 S,5 S)-2-Pinene
Type Analytical Standard
Quantity 1 mL
Formula Weight 136.23
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less