missing translation for 'onlineSavingsMsg'
Learn More

(-)-β-Pinene, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111019979
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019979 1 mL
111018435 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019979 Supplier Merck Emd Millipore Supplier No. 806091ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(-)-β-Pinene is a monoterpenoid, naturally found in the resins of pine trees and other conifers.It comprises of an exocyclic double bond in its structure.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Chemical Name or Material (-)-β-Pinene
CAS 18172-67-3
Flash Point 36°C
Physical Form Neat
Linear Formula C10H16
Technique Liquid Chromatography
MDL Number MFCD00001345
Boiling Point 164°C to 165°C
Synonym (1 S,5 S)-2(10)-Pinene; (1 S,5 S)-6,6-Dimethyl-2-methylenebicyclo[3.1.1 ]heptane
Type Analytical Standard
Molecular Weight (g/mol) 136.23
Quantity 1 mL
Formula Weight 136.23
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less