missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-α-Pinene, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 7785-70-8 |
| Ignition Point | 491°F |
| Density | 0.858 g/mL (at 20°C) |
| Flash Point | 33°C |
| Physical Form | Neat |
| Linear Formula | C10H16 |
| MDL Number | MFCD00001346 |
| Melting Point | -62°C |
| Boiling Point | 155°C to 156°C |
| Synonym | (1 R,5 R)-2,6,6-Trimethylbicyclo[3.1.1 ]hept-2-ene; (1 R,5 R)-2-Pinene |
| Show More |