Learn More
PIP™ Pinchfit™ BioSoft™ Ear Plug
Disposable, bio-based foam ear plug with Pinchfit™ Technology.
Supplier: PIP™ PF30
Description
- PIP™ understands the importance of reducing waste to lower costs and minimize environmental impact
- New, sustainable ECOSeries™ BioSoft™ ear plugs are made from bio-based technologies
- BioSoft™ ear plug foam material provides the same fit and performance as conventional polyurethane or PVC materials, but with a lower carbon footprint
- Innovative technology not only reduces emissions during manufacturing, but contains bio-based materials, making BioSoft™ more environmentally friendly at end of use
Specifications
| Yes | |
| Universal | |
| 1000 Plugs/Cs. | |
| Corded | |
| Single-use | |
| Yes | |
| Ear Plugs |
| Class AL | |
| No | |
| 32 NRR | |
| Bullet | |
| Yes | |
| EN 352-1 |