missing translation for 'onlineSavingsMsg'
Learn More

Pinacyanol Chloride 93.0+%, TCI America™

Catalog No. p-7134455
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0156100MG 100 mg
P01561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0156100MG Supplier TCI America Supplier No. P0156100MG

Chemical Identifiers

CAS 2768-90-3
Molecular Formula C25H25ClN2
Molecular Weight (g/mol) 388.94
MDL Number MFCD00011974
InChI Key FVMNARAKYNRZID-UHFFFAOYSA-M
Synonym pinacyanol chloride, quinaldine blue, unii-91sz6dgy86, vernitest, pinacyanochloride, vernitest reagent, 1,1'-diethyl-2,2'-carbocyanine chloride, 2,2'-trimethinequinocyanine chloride, chinaldinblau, 1-ethyl-2-3-1-ethylquinolin-1-ium-2-yl prop-2-enylidene quinoline chloride
PubChem CID 6364575
IUPAC Name 1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium chloride
SMILES [Cl-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12

Specifications

Melting Point 270°C
Quantity 100 mg
Formula Weight 388.94
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pinacyanol Chloride