accessibility menu, dialog, popup

UserName

ClCH3NNCH3(E/Z)(E/Z)

CAS RN 2768-90-3

IUPAC Name: 1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium chloride
Synonyms: pinacyanol(1+) chloride
Molecular Weight (g/mol): 388.94
Molecular Formula: C25H25ClN2
InChi Key: FVMNARAKYNRZID-UHFFFAOYSA-M
SMILES: [Cl-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results