missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinacyanol Chloride 93.0+%, TCI America™
Specifications
Specifications
| Melting Point | 270°C |
| Quantity | 1 g |
| Formula Weight | 388.94 |
| Percent Purity | ≥93.0% (HPLC) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pinacyanol Chloride |
Safety and Handling
| TSCA | Yes |