missing translation for 'onlineSavingsMsg'
Learn More

Pinacolin Oxime 98.0+%, TCI America™

Catalog No. P14735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14735G Supplier TCI America Supplier No. P14735G
Only null left

Chemical Identifiers

CAS 2475-93-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00019956
InChI Key UNSDDJQNHCSVSW-FNORWQNLSA-N
Synonym pinacolone oxime, 3,3-dimethyl-2-butanone oxime, 2-butanone,3-dimethyl-, oxime, 2-hydroxyimino-3,3-dimethylbutane, pinakolinoxim, 2e-n-hydroxy-3,3-dimethylbutan-2-imine
PubChem CID 9561079
IUPAC Name (NE)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
SMILES CC(=NO)C(C)(C)C

Specifications

Melting Point 76°C
Color White
Boiling Point 172°C
Quantity 5 g
Formula Weight 115.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pinacolin Oxime