missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinacol 98.0+%, TCI America™
$87.60 - $665.46
Chemical Identifiers
| CAS | 76-09-5 |
|---|---|
| Molecular Formula | C6H14O2 |
| Molecular Weight (g/mol) | 118.176 |
| MDL Number | MFCD00004462 |
| InChI Key | IVDFJHOHABJVEH-UHFFFAOYSA-N |
| Synonym | pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane |
| PubChem CID | 6425 |
| IUPAC Name | 2,3-dimethylbutane-2,3-diol |
| SMILES | CC(C)(C(C)(C)O)O |
Chemical Identifiers
| 76-09-5 | |
| 118.176 | |
| IVDFJHOHABJVEH-UHFFFAOYSA-N | |
| 6425 | |
| CC(C)(C(C)(C)O)O |
| C6H14O2 | |
| MFCD00004462 | |
| pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane | |
| 2,3-dimethylbutane-2,3-diol |
Specifications
| 76-09-5 | |
| White-Yellow | |
| C6H14O2 | |
| 25 g | |
| IVDFJHOHABJVEH-UHFFFAOYSA-N | |
| 2,3-dimethylbutane-2,3-diol | |
| 6425 | |
| ≥98.0% (GC) | |
| Pinacol |
| 43°C | |
| 174°C | |
| MFCD00004462 | |
| pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane | |
| CC(C)(C(C)(C)O)O | |
| 118.176 | |
| 118.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0233&(2)-0240
RTECSNumber : EK1720000
TSCA : Yes