missing translation for 'onlineSavingsMsg'
Learn More

Pinacol 98.0+%, TCI America™

Catalog No. D069125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D069125G 25 g
D0691100G 100 g
D0691500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D069125G Supplier TCI America Supplier No. D069125G

Chemical Identifiers

CAS 76-09-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004462
InChI Key IVDFJHOHABJVEH-UHFFFAOYSA-N
Synonym pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane
PubChem CID 6425
IUPAC Name 2,3-dimethylbutane-2,3-diol
SMILES CC(C)(C(C)(C)O)O

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 174°C
Quantity 25 g
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pinacol
EINECSNumber (2)-0233&(2)-0240
RTECSNumber EK1720000
TSCA Yes