missing translation for 'onlineSavingsMsg'
Learn More

Pimobendan, Thermo Scientific™

Catalog No. p-7232418
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468572500 250 mg
AC468570010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468572500 Supplier Thermo Scientific Chemicals Supplier No. 468572500
Only null left

Chemical Identifiers

CAS 74150-27-9
Molecular Formula C19H18N4O2
Molecular Weight (g/mol) 334.38
MDL Number 00761648
InChI Key GLBJJMFZWDBELO-UHFFFAOYNA-N
IUPAC Name 6-[2-(4-methoxyphenyl)-1H-1,3-benzodiazol-6-yl]-5-methyl-2,3,4,5-tetrahydropyridazin-3-one
SMILES COC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NNC(=O)CC1C

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Concentration or Composition Notes Titration with HClO4
Physical Form Powder
Concentration 97.5% to 102.5%
UN Number 2811
Color White to Light Yellow
Merck Index 7516
Quantity 250 mg
Formula Weight 334.37
Chemical Name or Material Pimobendan
Show More Show Less
Hazard Category GHS07: Exclamation mark
ShelfLife 3 years
RTECSNumber UR6373000
Recommended Storage Refrigerator +4°C