missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CG Mouse anti-Human, Clone:4B5D2, Abnova™
Mouse monoclonal antibody raised against recombinant human PIK3CG.
Supplier: Abnova Corporation MAB16608
Description
This gene encodes a protein that belongs to the pi3/pi4-kinase family of proteins. The gene product is an enzyme that phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring. It is an important modulator of extracellular signals, including those elicited by E-cadherin-mediated cell-cell adhesion, which plays an important role in maintenance of the structural and functional integrity of epithelia. In addition to its role in promoting assembly of adherens junctions, the protein is thought to play a pivotal role in the regulation of cytotoxicity in NK cells. The gene is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias. [provided by RefSeq]Specifications
| PIK3CG | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| PIK3CG | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 4B5D2 | |
| phosphoinositide-3-kinase, catalytic, gamma polypeptide | |
| In PBS (0.05% sodium azide) | |
| PI3CG, PI3K, PI3Kgamma, PIK3 | |
| Recombinant protein corresponding to amino acid 1-100 of human PIK3CG from E. coli. | |
| 5294 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |