missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3CA Recombinant Rabbit Monoclonal Antibody (HL4042)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA566202
Description
PIK3CA Recombinant Monoclonal Antibody for Western Blot
Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.Chemical Identifiers
| PIK3CA | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| P42336, P42337 | |
| 18706, 5290 | |
| Synthetic peptide of human PI3 kinase p110 alpha. | |
| Antibody |
| HL4042 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| PIK3CA | |
| 2.7.1.153; 6330412C24Rik; caPI3K; CLOVE; CWS5; EC:2.7.11.1; GRB1; MCAP; MCM; MCMTC; MGC142161; MGC142163; p110; p110alpha; p110-alpha; phophatidylinositol 3-kinase; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha; phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3K; PI3K alpha; PI3Kalpha; PI3K-alpha; pi3kca; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; Pik3ca; PIK3R1; pk3ca; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA | |
| PIK3CA | |
| Primary | |
| Protein A |
Specifications
| PIK3CA | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P42336, P42337 | |
| PIK3CA | |
| Synthetic peptide of human PI3 kinase p110 alpha. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| HL4042 | |
| Unconjugated | |
| PIK3CA | |
| 2.7.1.153; 6330412C24Rik; caPI3K; CLOVE; CWS5; EC:2.7.11.1; GRB1; MCAP; MCM; MCMTC; MGC142161; MGC142163; p110; p110alpha; p110-alpha; phophatidylinositol 3-kinase; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha; phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3K; PI3K alpha; PI3Kalpha; PI3K-alpha; pi3kca; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; Pik3ca; PIK3R1; pk3ca; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18706, 5290 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |