missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PIK3C3 (hVPS34), GST Tag Recombinant Protein
Recombinant Protein
$1,424.00 - $3,070.00
Specifications
| Accession Number | Q8NEB9 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Gene ID (Entrez) | 5289 |
| Molecular Weight (g/mol) | 129 kDa |
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
PIK3C3 (hVPS34) is the sole class III member of the PI3 Kinase family. PIK3C3 phosphorylates phosphatidylinositol (PI) to produce phosphatidylinositol 3-phosphate [PI(3)P], which recruits downstream effectors containing FYVE and Phox domains to membranes. PIK3C3 is involved in coupling nutrient signaling to mTOR and also plays a role in autophagy and intracellular trafficking. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.Specifications
| Q8NEB9 | |
| Kinase Assay | |
| 129 kDa | |
| VPS34 | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Liquid |
| See Label | |
| 5289 | |
| Human PIK3C3 (hVPS34), GST Tag | |
| PI3 Kinase Family | |
| Human | |
| GST-tag | |
| Baculovirus |