missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGW Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113078
Description
PIGW Polyclonal Antibody for Western Blot, ELISA
Phosphatidylinositol-glycans (PIGs) are multi-pass transmembrane proteins that localize to the endoplasmic reticulum. PIGs exhibit various functions, but all are crucial for the biosynthesis of the glycosylphosphatidylinositol (GPI)-anchor. Some PIG proteins are components of the GPI transamidase complex and play a role in the recognition of either the GPI attachment signal or the lipid portion of GPI. Other PIGs belong to the glycosyltransferase complex and function in the transfer of N-acetylglucosamine (GlcNAc) to phosphatidylinositol (PI). A variety of other PIGs play distinct roles in GPI synthesis including mannosylation of the GPI-anchor. PIG-W (Phosphatidylinositol-glycan biosynthesis class W protein) is a 504 amino acid multi-pass membrane protein that functions in the third step of GPI biosynthesis and acylates the inositol ring of phosphatidylinositol.Chemical Identifiers
| PIGW | |
| -20°C or -80°C if preferred | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| PIGW | |
| 2610044A17Rik; Gwt1; HPMRS5; phosphatidylinositol glycan anchor biosynthesis class W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; phosphatidylinositol-glycan biosynthesis class W protein; Pigw; PIG-W | |
| PIGW | |
| Primary | |
| Protein G |
| 3.7 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q7TSN4, Q7Z7B1 | |
| 284098, 378774 | |
| Recombinant Human Phosphatidylinositol-glycan biosynthesis class W protein (392-448AA). | |
| Antibody |
Specifications
| PIGW | |
| Polyclonal | |
| Unconjugated | |
| PIGW | |
| 2610044A17Rik; Gwt1; HPMRS5; phosphatidylinositol glycan anchor biosynthesis class W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; phosphatidylinositol-glycan biosynthesis class W protein; Pigw; PIG-W | |
| Rabbit | |
| Protein G | |
| RUO | |
| 284098, 378774 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.7 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q7TSN4, Q7Z7B1 | |
| PIGW | |
| Recombinant Human Phosphatidylinositol-glycan biosynthesis class W protein (392-448AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |