missing translation for 'onlineSavingsMsg'
Learn More

Pigment Red 53, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013523
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013523 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013523 Supplier Merck Emd Millipore Supplier No. 6719525MG

Specifications

CAS 2092-56-0
Physical Form Neat
Linear Formula C17H12 ClN2 NaO4 S
MDL Number MFCD00059530
Synonym 5-Chloro-2-[(2-hydroxy-1-naphthalenyl)azo]-4-methylbenzenesulfonic acid monosodium salt; Lake Red C
Type Analytical Standard
Quantity 25 mg
Formula Weight 398.80
Percent Purity ≥98% (HPLC)
Grade Analytical Standard