missing translation for 'onlineSavingsMsg'
Learn More

Pigment Blue 15, TCI America™

Supplier:  TCI America P065525G

Catalog No. P065525G

Only null left

Description

Description

Specifications

Specifications

Pigment Blue 15
147-14-8
MFCD00010719
576.08
25 g
480°C
C32H16CuN8
Phthalocyanine Blue, Copper Phthalocyanine, CuPc
Blue/Purple
Crystal/Powder

Chemical Identifiers

147-14-8
C32H16CuN8
MFCD00010719
Phthalocyanine Blue, Copper Phthalocyanine, CuPc
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3299&(5)-5216

RTECSNumber : GL8510000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions