missing translation for 'onlineSavingsMsg'
Learn More

New Pig Yellow Absorbent Mat Pads

Catalog No. 19033475
Change view
Click to view available options
Capacity (English):
28.16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19033475 28.16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19033475 Supplier New Pig Supplier No. MAT603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alert workers to spills and slip hazards while cleaning

  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Absorbs and retains oils, coolants, solvents and water
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Dispenser box makes it easy to take, carry and store your mat while protecting it from dirt and moisture
  • Color: yellow
  • Dimensions: 38 × 51cm (15 × 20 in.)

Specifications

Color Yellow
Capacity (English) 28.16 oz.
Capacity (Metric) 0.83 L
Material Polypropylene
Type High Visibility Absorbent Mat Pad
For Use With (Application) Absorbs oils, coolants, solvents and water
Dimensions (L x W) 20 x 15 in. (50.8 x 38 cm)
Length (English) 20 in.
Length (Metric) 50.8 cm
Width (English) 15 in.
Width (Metric) 38 cm
Weight (English) 17 lb.
Weight (Metric) 7.7 kg
Show More Show Less