missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, Crystal, Reagent Special, ACS, 99.95-105%, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Dichromate, Crystal, Reagent Special, ACS is an inorganic chemical reagent that is most often used as an oxidizing agent in industrial applications. It is known to be chronically detrimental to health. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 500 g |
| Percent Purity | 99.95 to 100.05% |
| Grade | ACS/Reagent |
| Loss on Drying | 0.05 |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Dichromate, Crystal |