missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Yeast Digest Standard

Catalog No. FERA47951
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA47951 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA47951 Supplier Thermo Scientific™ Supplier No. LSGA47951
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Thermo Scientific™ Pierce Yeast Digest Standard is a lyophilized yeast peptide mixture ideal for monitoring LC/MS system performance.

The Thermo Scientific™ Pierce Yeast Digest Standard is a lyophilized yeast peptide mixture ideal for monitoring LC/MS system performance.

Pierce Yeast Digest Standard features include:

  • Positive control sample – well-characterized eukaryotic proteome sample protein digest (>3,000 proteins) for LC/MS method standardization, development, and troubleshooting
  • Verified peptide quality – digestion conditions optimized for minimal missed cleavages or peptide modifications during processing
  • Rigorously tested – high-quality, consistent protein digest guaranteed and documented via lot-specific certificates of analysis
  • Convenient – ready-to-use lyophilized format

The Pierce Yeast Digest Standard is specifically formulated for LC/MS experiments and does not contain salts or detergents. The protein digest is derived from Saccharomyces cerevisiae yeast strain BY4741 (genetic background: MATa his3delta1 leu2delta0 met15delta0 ura3delta0). The yeast proteome is well-documented and characterized, and the digest is an ideal standard for more complex proteome mass spectrometry applications or when a non-mammalian digeste is required for analysis.

Using the standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.

Specifications

Chemical Name or Material Yeast peptide digest
Packaging Glass vial
Physical Form Lyophilized
Color White
Synonym Yeast peptide digest
Quantity 20 μg
Grade MS
Which LC/MS analysis process does Yeast Digest Standard help monitor?

Yeast Digest Standard is suitable for monitoring the Ion Source parameters (spray stability, source temperature of ionization, in-source fragmentation) and Mass Analyzer components (mass accuracy from scan to scan) of the LC/MS analysis.