missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vacuum Hydrolysis Tube, 6 mL

Catalog No. PI29571
Change view
Click to view available options
Quantity:
1 Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI29571 1 Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI29571 Supplier Thermo Scientific™ Supplier No. 29571

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality valved glass tubes designed for heated, vacuum, acid-hydrolysis of proteins and peptides for amino acid analysis by HPLC chromatography.

Thermo Scientific™ Pierce Vacuum Hydrolysis Tubes are high-quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis.

Highlights:

  • Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks

Specifications

Description Vacuum Hydrolysis Tube, 6mL
Product Type Vacuum Hydrolysis Tube
Content And Storage 0.0
Format Tube
Quantity 1 Tube

For Research Use Only. Not for use in diagnostic procedures.