missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ TMT11plex Yeast Digest Standard

Catalog No. PIA40939
Change view
Click to view available options
Quantity:
20 μg
5 x 20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA40939 5 x 20 μg
PIA40938 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA40939 Supplier Thermo Scientific™ Supplier No. A40939
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Pierce TMT11plex Yeast Digest Standard is a lyophilized yeast peptide mixture of four congenic yeast strains labeled with TMT11plex reagents to monitor LC-MS/MS system performance for multiplexed quantitation.

The Pierce TMT11plex Yeast Digest Standard consists of four congenic strains of Saccharomyces cerevisiae (a parental line, BY4741, and three lines lacking the non-essential proteins Met6, His4, or Ura2) labeled in triplicate (knock-out strains) or duplicate (parental strain) with TMT11-plex label reagents. The knock-out strains are used in combination with the parental strain to enable comparison of peptide and protein abundance for quantitation. Pierce TMT11plex Yeast Digest Standard:
• Convenient—ready-to-use lyophilized powder pre-labeled with TMT11plex reagents
• Quality—yeast strains verified for genotype, equimolar mixing of the TMT-labeled strains
• Quantitative—measure the quantitative precision of TMT reporter ions for 11 channels
• Diagnostic—excellent QC assay tool for protein quantitation using TMT on LC-MS/MS instruments

Quantitative proteomic strategies using TMT reagents enable multiplexing and precise measurement of protein abundance. Successful execution of TMT quantitation includes optimized chromatography, mass spectrometry (MS), and data analysis. A critical parameter for success in this workflow is the ability to consistently detect TMT fragment ions across 11 channels for multiplexing. Our TMT11plex Yeast Digest standard has been specifically designed to assess LC-MS/MS system performance for TMT quantitation, MS method optimization and system validation. For method development, TMT11plex Yeast Digest Standard can be used to measure the accuracy, precision, and dynamic range of different mass spectrometry approaches. These standards can help optimize offline fractionation, chromatography, mass spectrometry acquisition, and data analysis settings. Multifunctional applications using this standard reduces inter-experiment variability to provides reproducible TMT quantitation.

References:
1. Paulo JA, O'Connell JD, Gygi SP. A Triple Knockout (TKO) Proteomics Standard for Diagnosing Ion Interference in Isobaric Labeling Experiments. J Am Soc Mass Spectrom. 2016 10:1620-5.
2. Yang F, Shen Y, Camp DG 2nd, Smith RD. High-pH reversed-phase chromatography with fraction concatenation for 2D proteomic analysis. Expert Rev Proteomics. 2012 9(2):129-34.
3. Dayon L, Pasquarello C, Hoogland C, Sanchez JC, Scherl A. Combining low- and high-energy tandem mass spectra for optimized peptide quantification with isobaric tags. J Proteomics. 2010 73(4):769-77.
4. Diedrich JK, Pinto AF, Yates JR 3rd. Energy dependence of HCD on peptide fragmentation: stepped collisional energy finds the sweet spot. J Am Soc Mass Spectrom. 2013 24(11):1690-9.
5. Chiva C, Sabidó E. HCD-only fragmentation method balances peptide identification and quantitation of TMT-labeled samples in hybrid linear ion trap/orbitrap mass spectrometers. J Proteomics. 2014 96:263-70.

Related products:
Pierce HeLa Protein Digest Standard (Cat. No. 88329)
Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88321)
TMT10plex isobaric label reagent set plus TMT11-131C label reagent (Cat. No. A34808)

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Digest Standard
Product Line Pierce
Shipping Condition Wet Ice
Starting Material Peptides
Final Product Type MS Standard
Detection Method Mass Spectrometry
Workflow Step MS System Performance
Quantity 5 x 20 μg
Form Lyophilized Powder
After running the Pierce TMT11plex Yeast Digest Standard, I observe low signal to noise ratio and poor quantitation accuracy for the Tandem Mass Tag (TMT) reporter ions. What do you recommend?

Here are some recommendations:

- Clean and recalibrate the mass spectrometry instrument using one of our Pierce Calibration Solutions (https://www.thermofisher.com/search/browse/category/us/en/600654).
- Verify settings for liquid chromatography (LC) acquisition methods.
- Fractionate TMT-labeled samples using the Pierce High pH Reversed-Phase Peptide Fractionation Kit (Cat. No. 84868) to reduce sample complexity.

How do I use the Pierce TMT11plex Yeast Digest Standard to optimize my LC-MS acquisition parameters?

The Pierce TMT11plex Yeast Digest Standard (i.e., TKO standard) is a lyophilized yeast peptide mixture of four congenic strains labeled with TMT11plex reagents. It can be used to rapidly and accurately assess ion interference (co-isolation of multiple analytes of similar mass-to-charge) for TMT-based quantitative mass spectrometry proteomic analysis.

Which standard do you recommend using for Tandem Mass Tag (TMT) workflows?

We recommend using our Pierce TMT11plex Yeast Digest Standard (Cat. Nos. A40938, A40939) for LC-MS method optimization and for assessing system suitability for TMT-labeled samples.

Which standards and calibrants do you recommend using for assessing LC-MS system suitability?

Here are our recommendations:

- Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88321) for LC system and gradient optimization
- Pierce BSA Protein Digest, MS grade (Cat. No. 88341), Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342), and Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) for "bottom up" LC-MS method optimization and LC-MS suitability assessment
- Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) (Cat. No. A40010) for assessing the gradient, sensitivity, and dynamic range of the LC-MS system for discovery and targeted peptide quantitative workflows
- Pierce TMT11plex Yeast Digest Standard (Cat. Nos. A40938, A40939) for assessing system performance for TMT-based quantitative workflows
- Pierce Intact Protein Standard Mix (Cat. Nos. A33526, A33527) for "top down" LC-MS method optimization and LC-MS suitability assessment

What type of mass spectrometer can I use for Tandem Mass Tag (TMT) analysis?

We recommend using high-resolution Orbitrap Tribrid (e.g., Fusion, Lumos, Eclipse), Orbitrap Exploris (e.g., 240, 480), or Q Exactive (e.g,. Plus, HF, HFX) series of instruments.

What is the Pierce TMT11plex Yeast Digest Standard and what is its main purpose?

The Pierce TMT11plex Yeast Digest Standard is a lyophilized yeast peptide mixture of four congenic strains labeled with TMT11plex reagents. It serves as a standard for quality assessment of LC and MS data for samples labeled with Tandem Mass Tag (TMT) reagents.


For Research Use Only. Not for use in diagnostic procedures.