missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent)

Catalog No. pi27712
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI27712 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI27712 Supplier Thermo Scientific™ Supplier No. LSG27712

Add tyrosine (tyrosyl) groups to proteins to increase the number of iodine-125 labeling (iodination) sites using an amine-reactive, NHS ester compound.

Thermo Scientific™ Pierce Sulfo-SHPP (water-soluble Bolton-Hunter Reagent) conjugates tyrosine-like functional groups to primary amines to increase the number of tyrosyl groups that can be iodinated by iodine-125 labeling procedures.

Highlights:

  • Membrane-impermeable
  • Ideal for selectively radiolabeling cell surface proteins

Specifications

Product Type Sulfo-SHPP
Quantity 100 mg
Content And Storage Upon receipt store at -20°C. Product is shipped at ambient temperature.
Product Line Pierce
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Iodine-125
Spacer Short
Label Type Radioisotope
Show More Show Less
Can you provide the shelf-life for Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent)?

Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.