missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Kit - Western Blot Application

Catalog No. PI33073
Change view
Click to view available options
Quantity:
8 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI33073 8 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI33073 Supplier Thermo Scientific™ Supplier No. 33073

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tag a purified protein to covalently crosslink and transfer biotinylation to its binding partner; then detect the results by Western blotting.

Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer WB Kit includes Sulfo-SBED Reagent to crosslink interacting proteins and transfer a biotin label from starting protein to target protein for detection by Western blotting.

Label transfer is a powerful in vitro method for protein interaction discovery. Sulfo-SBED Biotin Label Transfer Reagent can be used to identify previously unknown protein interaction binding partners and to more fully characterize the specific protein binding domains of other protein interactions. Sulfo-SBED is the abbreviation for Sulfo-N-hydroxysuccinimidyl-2-(6-[biotinamido]-2-(p-azido benzamido)-hexanoamido) ethyl-1,3'-dithioproprionate. It is a heterobifunctional chemical crosslinker capable of covalently attaching to primary amines at one end and to nearly any protein functional group at the other end. Unlike typical crosslinkers, Sulfo-SBED also includes a biotin group and a cleavable disulfide spacer arm. Together these features allow one to sequentially crosslink interacting proteins and transfer the biotin affinity tag from one protein (i.e., a purified “bait” protein) to another (possibly unknown “prey” protein).

Highlights:

  • Sulfo-NHS ester reacts with primary amines at pH 7-9
  • Aryl azide conjugates randomly when activated with UV light
  • Cleavable disulfide bond allows biotin label transfer
  • Discover protein interactions without radiolabeling

Includes:

Sulfo-SBED No-Weigh™ Format 8 x 1mg microtubes, BupH™ Phosphate Buffered Saline 1 pack, Label Transfer Buffer (20X) 200mL, Streptavidin-Horseradish Peroxidase Conjugate 0.1mg, Dithiothreitol (DTT) No-Weigh Format 8 x 7.7mg microtubes, Slide-A-Lyzer™ MINI Dialysis Units Plus Float 10kDa MWCO 10 units/pack

Specifications

Format Kit
Quantity 8 Reactions
Product Type Crosslinker
Solubility Water
Content And Storage Sufficient For: 8 label transfer reactions for subsequent western blot analysis
• Sulfo-SBED, 8 x 1 mg
• PBS pack (makes 500 mL), 1 pack
• Label Transfer Buffer (20X), 200 mL
• Streptavidin-HRP Conjugate, 0.1 mg
• Dithiothreitol (DTT), 8 x 7.7 mg
• Slide-A-Lyzer MINI Dialysis Devices Plus Float, 10K MWCO, 10-device kit

Store at 4°C
Labeling Method Chemical Labeling, Label Transfer
Reactive Moiety Sulfo-NHS Ester, Aryl Azide
Product Line Pierce
What are the label transfer reagents you offer?

Currently, we offer sulfo-SBED Biotin Label Transfer Reagent (Cat. Nos. 33033 and 33034) as well as the Sulfo-SBED Biotin Label Transfer Kit for Western Blot Application (Cat. No. 33073).

For Research Use Only. Not for use in diagnostic procedures.