missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Poly-HRP

Catalog No. PI21140
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21140 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21140 Supplier Thermo Scientific™ Supplier No. 21140
  • ONLINE EXCLUSIVE PRICE
    $667.34
    $567.24 / Each

    Save $100.10

    15.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amplify signal and improve detection of biotinylated antibodies for IHC and other methods using this streptavidin conjugated with HRP polymers.

Thermo Scientific™ Pierce Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods.

This poly-HRP conjugate is designed to deliver the highest sensitivity and low background in immunoassays where sample volume is limited or when the target molecule is present at low levels. Pierce Streptavidin Poly-HRP is purified to remove unconjugated streptavidin molecules that reduce signal intensity by competing for binding sites with horseradish peroxidase (HRP)-conjugates molecules. In addition, the conjugate is devoid of HRP monomers that can cause background signal. The Pierce Streptavidin Poly-HRP is compatible with chromogenic, fluorogenic and chemiluminescent HRP substrates used in ELISA, Western blotting, immunohistochemistry (IHC) and nucleic acid hybridization assays.

Highlights:

  • High sensitivity – detect low-abundant targets (low picogram-femtogram range) with high signal-to-noise ratios

  • Robust – consistent manufacturing and purification to eliminate low- and un-conjugated molecules for minimal background and high sensitivity

  • Flexible – compatible with chromogenic, fluorogenic and chemiluminescent substrates

  • Versatile – compatible with ELISA, Western blotting and immunohistochemistry (IHC)

  • Easy to use – can be directly substituted into immunoassays and other detection assays

  • Convenient – ready-to-use stabilized liquid format stored at 4°C

  • Cost effective – requires less conjugate per assay and standard HRP conjugates

Specifications

Content And Storage Stored at 4°C
Specificity Biotin
Enzyme HRP (Horseradish Peroxidase)
Form Liquid
For Use With (Application) Western Blot, ELISA, Immunohistochemistry
Product Line Pierce
Label or Dye HRP (Horseradish Peroxidase)
Target Molecule Biotin
Quantity 0.5 mL

For Research Use Only. Not for use in diagnostic procedures.