missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ High Sensitivity Streptavidin-HRP
Description
Thermo Scientific™ Pierce High Sensitivity Streptavidin HRP Conjugate is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
- Thermo Scientific Pierce High Sensitivity Streptavidin-HRP is an exclusive peroxidase-conjugated streptavidin biotin-binding protein
- It provides signal amplification like poly-HRP and exceptional storage stability
- This specially-manufactured variety of HRP-conjugated streptavidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications
- Conjugate is suitable for use with chemiluminescent, chemifluorescent, or colorimetric substrates
- Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year
- Experiments indicate that the High Sensitivity Streptavidin-HRP can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results
Specifications
Specifications
| Concentration | 1.0–1.1 mg/mL |
| Conjugate | HRP |
| Quantity | 5 mL |
| Content And Storage | Storage at 4°C for at least one year. |
| Product Line | Pierce |
| Target Molecule | Biotin |
For Research Use Only. Not for use in diagnostic procedures.