missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Sensitivity Streptavidin-HRP

Catalog No. p-4532004
Change view
Click to view available options
Quantity:
0.5 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21130 0.5 mL
PI21132 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
d
Catalog No. PI21130 Supplier Thermo Scientific™ Supplier No. LSG21130
Only null left
d

Amplify signal in Western blot, ELISA or other biotin-based detection methods with this exclusive, high signal-to-noise, peroxidase-conjugated, ready-to-use streptavidin.

Thermo Scientific™ Pierce High Sensitivity Streptavidin HRP Conjugate is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

  • Thermo Scientific Pierce High Sensitivity Streptavidin-HRP is an exclusive peroxidase-conjugated streptavidin biotin-binding protein
  • It provides signal amplification like poly-HRP and exceptional storage stability
  • This specially-manufactured variety of HRP-conjugated streptavidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications
  • Conjugate is suitable for use with chemiluminescent, chemifluorescent, or colorimetric substrates
  • Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year
  • Experiments indicate that the High Sensitivity Streptavidin-HRP can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results

Specifications

Concentration 1.0–1.1 mg/mL
Conjugate HRP
Detection Method Colorimetric, Chemiluminescence, Chemifluorescence
Quantity 0.5 mL
Content And Storage Storage at 4°C for at least one year.
Form Liquid
Product Line Pierce
For Use With (Application) Western Blotting, ELISA
Specificity Biotin
Target Molecule Biotin

For Research Use Only. Not for use in diagnostic procedures.