missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin

Catalog No. PI21122
Change view
Click to view available options
Quantity:
1 mg
5 mg
100 mg
1000 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21122 1 mg
PI21125 5 mg
PI21135 100 mg
PI21145 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI21122 Supplier Thermo Scientific™ Supplier No. 21122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.

Thermo Scientific Pierce Streptavidin is a purified unconjugated recombinant protein form.

Features of Streptavidin:

  • Recombinant streptavidin—high-quality protein, 53kDa, pI = 6.8 to 7.5
  • Pure protein —choose pure, unconjugated protein for biotin blocking and controls

The purified recombinant Streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including Western blotting, ELISA, immunohistochemistry and fluorescence imaging. Purified streptavidin is supplied as lyophilized in phosphate-based buffers for immediate reconstitution with water.

Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific Pierce Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.

Related Products

  • Pierce™ Streptavidin, Horseradish Peroxidase Conjugated (Cat. No. 21126)
  • Pierce™ Streptavidin, Alkaline Phosphatase Conjugated (Cat. No. 21324)
  • Pierce™ Streptavidin, Maleimide-Activated (Cat. No. 21102)
  • Pierce™ Streptavidin, Hydrazide-Activated (Cat. No. 21120)

Specifications

Conjugate Unconjugated
Quantity 1 mg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Form Solid
Product Line Pierce
Product Type Streptavidin
Target Molecule Biotin
Label or Dye Unlabeled
How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

For Research Use Only. Not for use in diagnostic procedures.