Learn More
Thermo Scientific™ Pierce™ Saturated Ammonium Sulfate Solution
Fractionate immunoglobulin, antibody, or other proteins by selective precipitation, also called salting out.
Supplier:  Thermo Scientific™ 45216
Catalog No. PIA45216
Only null left
Explore available promotions
Description
Description
Thermo Scientific™ Pierce Saturated Ammonium Sulfate Solution is formulated and ready to use for simple immunoglobulin, antibody, or other protein fractionation applications based on selective precipitation, which is also called salting out.
The saturated ammonium sulfate solution (4.32M) enables removal of abundant proteins from serum, cell culture supernatant and ascites fluid. Slowly adding the solution to a protein sample results in decreased protein solubility and eventual protein precipitation as the charged ammonium sulfate ions compete with protein for water interaction. High molecular weight proteins, such as antibodies, are among the first to precipitate. When used in combination with antibody purification methods such as affinity chromatography, ammonium sulfate precipitation is ideal for achieving highly purified antibody samples.
Highlights:
  • Convenient – solution is ready to use; no preparation required
  • Concentrates – recover antibody in 100mL from more than 1L of cell culture supernatant
  • Improves purity – partial-purification of antibodies by ammonium sulfate precipitation greatly improves the final purity that can be obtained with affinity purification
  • Extends column life – using pre-purified antibody samples instead of crude samples enables affinity columns to be used for many more cycles
  • Compatible – ammonium sulfate is easily removed by dialysis or desalting columns, eliminating any obstacles to downstream methods
  • Versatile – use for precipitating immunoglobulins or “salting out” fractions of other sample types
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Specifications
Ammonium Sulfate Solution
Affinity
Upon receipt store at ambient temperature.
Liquid
Pierce™
Precipitation Reagent
1 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.