Learn More
Thermo Scientific™ Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Thermo Scientific Pierce S-Nitrosylation Western Blot Kit enables sensitive detection of protein S-nitrosocysteine post-translational modifications in a complete, easy-to-use kit or stand-alone HENS buffer for S-Nitrosylation Western Blot Kit.
$311.90 - $1,197.48
Specifications
| Detection Method | S-Nitrosylation |
|---|---|
| Product Line | Pierce |
| Reactivity | All species |
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
|
PI90105
|
Thermo Scientific™
LSG90105 |
40 Reactions | Western Blot Kit |
Each for $1,197.48
|
|
||||
|
PI90106
|
Thermo Scientific™
90106 |
100 mL | HENS Buffer |
Each for $311.90
|
|
||||
Description
The Pierce S-Nitrosylation Western Blot Kit provides cell lysis/reaction buffer, sulfhydryl-reactive blocking agent, reducing agent, labeling agent, and anti-TMT antibody to label 40 samples (100 μg each). As with traditional S-nitrosylation switch assays, unmodified cysteines are blocked by a sulfhydryl-reactive compound. S-nitrosylated cysteines are selectively reduced with ascorbate in HENS Buffer for labeling with iodoTMTzero reagents, which irreversibly bind to the cysteine thiol that was S-nitrosylated. Detection of the TMT reagent-modified proteins is facilitated using an anti-TMT antibody.
S-nitrosylation is a post-translational modification that regulates numerous processes, including cellular proliferation, apoptosis, smooth muscle relaxation, neurotransmitter release, and differentiation. During S-nitrosylation, nitric oxide radicals react with cysteine thiols to produce an S-NO adduct that alters protein activity.Pierce S-Nitrosylation Western Blot Kit
The Pierce S-Nitrosylation Western Blot Kit provides the necessary reagents to perform a modified version of the traditional S-nitrosylation switch assay, including a cysteine-blocking reagent, reducing agent, reaction buffer, iodoTMT labeling reagent, and anti-TMT antibody. Labeling is accomplished using the non-biological iodoTMT reagent instead of HPDP-biotin, reducing background signal during western blot detection. In addition to detection of S-nitrosylated proteins by western blot, an immobilized anti-TMT resin can be used to selectively enrich S-nitrosylated proteins/peptides labeled with iodoTMT reagents. This workflow allows for S-nitrosylation site mapping and multiplexed quantitation using mass spectrometry.
Features of Pierce S-Nitrosylation Western Blot Kit include:
• Specific—optimized labeling reactions switch S-nitrosocysteine post translational modifications with iodoTMT reagent for easy detection by western blot using anti-TMT antibody
• Better signal-to-noise—uses a non-biological iodoTMT reagent for labeling instead of HPDP-biotin, resulting in less background during western blot detection
• Workflow-compatible—labeling method is compatible with alternative methods of analysis, including enrichment via anti-TMT resin and multiplex quantitation by mass spectrometry
• Convenient—all needed reagents included one kit
HENS Buffer
HENS Buffer consists of 100 mM HEPES (pH 7.8), 1 mM EDTA, 0.1 mM Neocuproine, and 1% SDS. It is a cell lysis buffer intended for use with the Pierce S-Nitrosylation Western Blot Kit, which enables sensitive detection of protein S-nitrosocysteine post-translational modifications in a complete, easy-to-use kit.
Specifications
| S-Nitrosylation | |
| Pierce | |
| All species |