Learn More
Thermo Scientific™ Pierce™ Renilla Luciferase Glow Assay Kit
Description
Obtain bright and stable bioluminescence signal (half-life approx. 3 hour) in the presence of Renilla luciferase reporters.
Thermo Scientific™ Pierce Renilla Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life approx. 3 hours), especially in the presence of Green Renilla luciferase reporter.
This Pierce Luciferase Glow Assay Kit contains reagents for measuring the activity of Renilla luciferase in cell lysates. In the presence of luciferase, the glow reagent provides stable bioluminescent signal and is highly suited for luminometers without injectors or for batch processing of samples. The light output generated by the luciferase reaction can be correlated with the amount of luciferase protein produced which in turn is proportional to the promoter activity driving the luciferase expression. The glow assay reagents were optimized for best results with Thermo Scientific Green Renilla Luc Plasmids; however, this glow kit may be used to detect the activity of other Renilla luciferase derivatives that use coelenterazine as a substrate.
Highlights
- Sensitive—highly sensitive detection of Green Renilla luciferase activity
- Stable—increased signal stability compared to flash assays
- Simple—does not require a luminometer with injectors
- Compatible—assay reagents compatible with other Renilla luciferases
- Automation-friendly—amenable to high throughput assays
- Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
- Safe—allows one to perform non-radioactive assays
Requires
Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers
Recommended applications
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Green Renilla Luciferase |
| Quantity | 100 Reaction Kit |
| Content And Storage | Upon receipt store kit at -80°C or store individual components as indicated above. • Renilla Glow Assay Buffer, 5 mL, store at 4°C • Coelenterazine (100X), 50 μL, store at -80°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Label Type | Enzyme Labeled |
| Show More |
Frequently Asked Questions (FAQs)
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Use new sample; Change pipette tips after each well.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.
Here are possible causes and solutions:
- Non-optimized lysis buffer used: Assay luciferase activity in the media to confirm good expression of luciferase; Use only the provided lysis buffer.
- Low luciferase expression: Lyse cells in smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale up volume of sample and reagent per well.
Here are possible causes and solutions:
- Insufficient luciferase accumulation in media: Incubate cells for a longer time.
- Low luciferase expression: Use less media per well during the experiment;.Use a different promoter or growth conditions to improve expression;.Increase the integration time on the instrument;.Scale-up the volume of sample and reagent per well.
- Treatment interfered with cellular secretory pathway: Transfect cells with a plasmid for constitutive expression of Luciferase (i.e., with pTK or pCMV promoter); Determine if luciferase actively expresses in media without treatment. Add treatment; Determine if there is a corresponding drop in luciferase activity from the constitutively expressed plasmid.
Here are possible causes and solutions:
- Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality - use only transfection grade DNA; Use actively dividing, low passage cells; Use a different cell type.
- No or low promoter activity: Use conditions known for promoter activation; Incubate cells for a longer time; Change growth conditions to improve expression; Use a different promoter.
- Substrate auto-oxidized: Protect substrate from light and air; Maintain 100X Coelenterazine at -80 degrees C; maintain 100X Vargulin at -80 degrees C; Prepare new Coelenterazine Working Solution if used longer than 8 hours; Prepare new Vargulin Working Solution if used longer than 2 hours
For Research Use Only. Not for use in diagnostic procedures.