Learn More
Thermo Scientific™ Pierce™ Renilla Luciferase Flash Assay Kit
Description
Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based Renilla luciferase kit.
The Thermo Scientific™ Pierce Renilla Luciferase Flash Assay Kit provides researchers with a highly sensitive intracellular assay for transcriptional activity of regulatory elements in mammalian whole cell lysate.
This Flash Assay Kit contains reagents for measuring the activity of Renilla luciferase in mammalian cell lysates. When used with Thermo Scientific Green Renilla Luc Plasmids, the kit provides an extremely sensitive bioluminescent reporter assay system for intracellular detection of promoter or pathway activity. The signal produced by Green Renilla Luc is considerably greater than signal from either native firefly or Renilla luciferases assayed under similar conditions. Light output captured using a luminometer can be correlated with the amount of Renilla luciferase protein produced and used to determine the activity of the promoter driving Renilla expression.
Highlights
- Sensitive—greater sensitivity allows utilization of smaller numbers of cells
- Cost effective—highest sensitivity assays result in decreased reagent consumption
- Time-saving—assays require minimal sample handling
- Automation-friendly—amenable to high throughput screens
- Convenient—contains a universal cell lysis buffer and optimized flash assay reagent
- Safe—allows one to perform non-radioactive assays
Requires
Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers
Recommended applications
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Green Renilla Luciferase |
| Quantity | 100 Reaction Kit |
| Content And Storage | • Renilla Flash Assay Buffer, 5 mL, store at 4°C • Coelenterazine (100X), 50 μL, store at -80°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Label Type | Enzyme Labeled |
| Show More |
Frequently Asked Questions (FAQs)
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Use new sample; Change pipette tips after each well.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.
Here are possible causes and solutions:
- Non-optimized lysis buffer used: Assay luciferase activity in the media to confirm good expression of luciferase; Use only the provided lysis buffer.
- Low luciferase expression: Lyse cells in smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale up volume of sample and reagent per well.
Here are possible causes and solutions:
- Insufficient luciferase accumulation in media: Incubate cells for a longer time.
- Low luciferase expression: Use less media per well during the experiment;.Use a different promoter or growth conditions to improve expression;.Increase the integration time on the instrument;.Scale-up the volume of sample and reagent per well.
- Treatment interfered with cellular secretory pathway: Transfect cells with a plasmid for constitutive expression of Luciferase (i.e., with pTK or pCMV promoter); Determine if luciferase actively expresses in media without treatment. Add treatment; Determine if there is a corresponding drop in luciferase activity from the constitutively expressed plasmid.
Here are possible causes and solutions:
- Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality - use only transfection grade DNA; Use actively dividing, low passage cells; Use a different cell type.
- No or low promoter activity: Use conditions known for promoter activation; Incubate cells for a longer time; Change growth conditions to improve expression; Use a different promoter.
- Substrate auto-oxidized: Protect substrate from light and air; Maintain 100X Coelenterazine at -80 degrees C; maintain 100X Vargulin at -80 degrees C; Prepare new Coelenterazine Working Solution if used longer than 8 hours; Prepare new Vargulin Working Solution if used longer than 2 hours
For Research Use Only. Not for use in diagnostic procedures.