Learn More
Thermo Scientific™ Pierce™ Renilla-Firefly Luciferase Dual Assay Kit
Description
Simultaneously detect intracellular Renilla and Red Firefly luciferase activity in mammalian whole cell lysates.
The Thermo Scientific™ Pierce Renilla-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Renilla and Red Firefly luciferase activity in mammalian whole cell lysates.
The Pierce Renilla-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Green Renilla and Red Firefly luciferase activity. Green Renilla luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. This reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching.
The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.
Highlights
- Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
- Sensitive—measure Green Renilla and red firefly luciferase activities in the same sample
- Fast—no quenching step required, unlike in traditional sequential dual assays
- Multiplex—capable of quantitating two cellular activities in the same sample(s)
Requires
Green Renilla and Red Firefly luciferase reporters; filter sets at 525 nm±20 nm for Green Renilla Luc and 615 to 675 nm for Red Firefly Luc; luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; injector for assessing more than 24 wells at a time
Recommended applications
Study two regulatory elements at the same time; monitor two signaling pathways simultaneously; enable studying more than one target per screen (e.g., off-target effects)
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Firefly Luciferase, Green Renilla Luciferase |
| Sufficient For | 100 Dual Luciferase Assays |
| Quantity | 100 Reaction Kit |
| Content And Storage | Sufficient For: 100 dual-luciferase assays in microplate wells • Renilla-Firefly Dual Assay Buffer, 5 mL (store at 4°C) • 100X Coelenterazine, 50 μL (store at -80°C) • D-Luciferin, 3 mg (store at 4°C) • Luciferase Cell Lysis Buffer (2X), 6 mL (store at room temperature) Upon receipt, store kit at -80°C, or store individual components as indicated above. |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Show More |
Frequently Asked Questions (FAQs)
The Pierce Renilla-Firefly Luciferase Dual Assay Kit (Cat. Nos. 16185, 16186) luciferase vectors were optimized with TurboFect. However, it should be feasible to replace TurboFect with another transfection reagent if your cells tolerate the transfection reagent better and give you acceptable transfection rates.
The stability of the Pierce Renilla-Firefly Luciferase Dual Assay Kit (Cat. Nos. 16185, 16186) may be inconsistent from sample to sample at -20 degrees C. We recommend snap-freezing lysates at -80 degrees C with protease inhibitors to help prevent protein degradation.
No. All Luciferase Dual Assay Kits, such as the Pierce Renilla-Firefly Luciferase Dual Assay Kit (Cat. Nos. 16185, 16186), are flash-based assays. Flash luminescence reactions give a very bright signal over a short reaction time. Luciferase assays that are flash-based require the use of a luminometer or other luminescence-monitoring instrument equipped with automated reagent injectors.
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use a new sample; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Change pipette tips after each well; Reduce shaker speed during the cell lysis step to avoid contaminating the wells.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.
For Research Use Only. Not for use in diagnostic procedures.