missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Recombinant Protein L
Detect mouse and human antibodies containing kappa light chains in immunoassays using this purified, biotinylated or peroxidase-conjugated, recombinant Protein L.
$273.36 - $2,059.79
Specifications
| Product Line | Pierce |
|---|---|
| Form | Powder |
| Protein Subtype | Protein L |
| Target | Antibodies |
| Content And Storage | Upon receipt store at 4°C. Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C. |
Description
Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein L enable probing and detection of mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.
Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.Highlights:
- Contains four immunoglobulin-binding domains per protein
- Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
- Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
Specifications
| Pierce | |
| Protein L | |
| Upon receipt store at 4°C. Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C. |
| Powder | |
| Antibodies |
For Research Use Only. Not for use in diagnostic procedures.