missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein A Agarose

Catalog No. PI20366
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20366 25 mL
PI20365 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20366 Supplier Thermo Scientific™ Supplier No. 20366

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purify IgG using this genetically engineered, immobilized protein A on beaded agarose, when native protein A has undesirable effects.

Thermo Scientific™ Pierce Recombinant Protein A Agarose is a beaded agarose resin to which a genetically engineered form of Protein A has been immobilized for use in IgG purification applications where the native ligand has undesirable effects.

The Pierce Recombinant Protein A Agarose uses a genetically engineered form of Protein A that is produced in E. coli and then immobilized on crosslinked 6% beaded agarose (CL-6B). Although some nonessential regions have been removed, recombinant is similar to native Protein A in overall size and IgG-binding domains. The protein has four high affinity binding sites capable of specifically binding to the Fc region of immunoglobulin molecules from several species. Some researchers prefer to use an affinity resin based on the recombinant form of Protein A in particular applications where practical experience indicates that the different production source (E. coli vs. S. aureus) provides less nonspecific binding or lower levels of endotoxin contamination. Otherwise, native Protein A resins generally provide higher capacity antibody purification.

Highlights:

  • Immobilized Protein A – specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
  • Standard capacity – loading of immobilized Protein A provides a binding capacity of 15 to 17mg human IgG per milliliter of resin
  • Agarose resin – crosslinked 6% beaded agarose for small column or batch affinity purification methods

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Specifications

Product Type Agarose
Product Line Pierce
Column Type Agarose Resin, Affinity
Format Bottle
Stationary Phase Protein A
Purification Target IgG Antibodies
Content And Storage Upon receipt store product at 4°C to 8°C. Product is shipped at ambient temperature.
Quantity 25 mL

For Research Use Only. Not for use in diagnostic procedures.