Learn More
Thermo Scientific™ ELISA Reagent Reservoir
Catalog No. PI15075
Change view
Click to view available options
Quantity:
200 Units
Catalog No. Quantity
PI15075 200 Units
1 options
Catalog No. PI15075 Supplier Thermo Scientific™ Supplier No. 15075
Rapidly dispense reagents with multi-channel pipettors using these sterile, sturdy and disposable reservoirs molded from high-impact polystyrene.
Thermo Scientific™ Pierce Reagent Reservoirs are sterile, sturdy, disposable and molded from high-impact polystyrene.
Additional Information and Online Ordering
Reservoirs facilitate repeated multi-channel pipetting for reagent delivery to microplates.
Highlights:
  • Sterile, sturdy reservoirs are molded from high-impact polystyrene
    Reservoirs facilitate repeated pick up of reagents by multi-channel pipettors for delivery to 96-well plates
Specifications
Description Pierce ELISA Reagent Reservoirs
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Quantity 200 Units
Content And Storage Store at room temperature.
Plate Compatibility 96-Well Plate, 384-Well Plate
Disposable Single-use
Bottom Shape V-bottom
For Research Use Only. Not for use in diagnostic procedures.