missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein G UltraLink™ Resin

Extremely durable polyacrylamide support with immobilized, recombinant Protein G for various IgG purification methods.

Supplier:  Thermo Scientific™ 53126

 View more versions of this product

Catalog No. PI53126


Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce Protein G UltraLink Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein G has been immobilized for use in IgG purification methods.

The Protein G UltraLink Resin is used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein G is a popular antibody-binding protein that is especially useful for affinity purification of IgG from mouse, human and rat serum.

UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.

Highlights:

  • Immobilized Protein G – specifically binds IgG from several mammalian species, especially all isotypes of IgG from human, mouse and rat

  • Standard capacity – loading of immobilized Protein G provides a binding capacity of approx. 20mg human IgG per milliliter of resin

  • UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications

Properties of Thermo Scientific UltraLink Resin:

  • Support pH Stability: 1 to 13
  • Average Particle Size: 50 to 80 microns
  • Exclusion Limit: greater than 2,000,000 daltons
  • Average Surface Area: greater than 250m2 per gram of beads
  • Average Pore Volume: greater than 1.2mL per gram of beads (> 60% of bead volume)
  • Pore Size: 500 to 1000 angstroms (50 to 100nm)
  • Maximum Linear Velocity: 3000cm per hour
  • Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
Specifications

Specifications

Resin
UltraLink Affinity Resin
Protein G
Upon receipt store product at 4°C. Products are shipped at ambient temperature.
Pierce, UltraLink
Bottle
IgG Antibodies
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.