missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A/G UltraLink™ Resin

Catalog No. PI53133
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI53133 10 mL
PI53132 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI53133 Supplier Thermo Scientific™ Supplier No. 53133

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extremely durable polyacrylamide support with immobilized recombinant Protein A/G for various IgG purification methods.

Thermo Scientific™ Pierce Protein A/G UltraLink Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein A/G has been immobilized for use in IgG purification methods.

The Protein A/G UltraLink Resin is used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein A/G is a recombinant fusion protein consisting of the antibody-binding domains of Protein A and Protein G, enabling antibody purification from serum samples of a variety of species.

UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.

Highlights:

  • Immobilized Protein A/G – specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG

  • Standard capacity – loading of immobilized Protein A/G provides a binding capacity of approx. 20mg human IgG per milliliter of resin

  • UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications

Properties of Thermo Scientific UltraLink Resin:

  • Support pH Stability: 1 to 13
  • Average Particle Size: 50 to 80 microns
  • Exclusion Limit: greater than 2,000,000 daltons
  • Average Surface Area: greater than 250m2 per gram of beads
  • Average Pore Volume: greater than 1.2mL per gram of beads (> 60% of bead volume)
  • Pore Size: 500 to 1000 angstroms (50 to 100nm)
  • Maximum Linear Velocity: 3000cm per hour
  • Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Specifications

Product Type Resin
Product Line Pierce, UltraLink
Column Type UltraLink Affinity Resin
Format Bottle
Stationary Phase Protein A/G
Purification Target IgG Antibodies
Content And Storage Upon receipt store resin at 4°C. Product is shipped at ambient temperature.
Quantity 10 mL

For Research Use Only. Not for use in diagnostic procedures.