missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A/G Plus Agarose

Supplier:  Thermo Scientific™ 20423

 View more versions of this product

Catalog No. PI20423


Only null left
Explore available promotions

Description

Description

Pierce Protein A/G Plus Agarose consists of purified Protein A/G recombinant fusion protein that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). Because the Protein A/G combines the immunoglobulin-binding domains of both Protein A and Protein G, the resin is effective for affinity purification of IgG antibodies from a broad range mammalian host species.

Highlights:

  • Protein A/G – immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species

  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods

  • Inert and stable – superior manufacturing method immobilizes Protein A/G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield

  • High capacity – “Plus” variety of Pierce Protein A/G Agarose has a very dense load of immobilized Protein A/G, providing a binding capacity greater than 50mg human IgG/mL resin

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
Specifications

Specifications

Agarose
Agarose Resin, Affinity
Protein A/G
Upon receipt store product at 4°C. Product is shipped with wet ice pack.
Pierce
Bottle
IgG Antibodies
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.