missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A/G Plus Agarose

Catalog No. PI20424
Change view
Click to view available options
Quantity:
2 mL
20 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20424 20 mL
PI20423 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20424 Supplier Thermo Scientific™ Supplier No. 20424

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Pierce Protein A/G Plus Agarose is an exceptionally high-capacity Protein A/G beaded agarose resin for use in a variety of antibody affinity purification methods.

Thermo Scientific Pierce Protein A/G Plus Agarose is an exceptionally high-capacity Protein A/G beaded agarose resin for use in a variety of antibody affinity purification methods.

Pierce Protein A/G Plus Agarose consists of purified Protein A/G recombinant fusion protein that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). Because the Protein A/G combines the immunoglobulin-binding domains of both Protein A and Protein G, the resin is effective for affinity purification of IgG antibodies from a broad range mammalian host species.

Highlights:

  • Protein A/G – immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species
  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable – superior manufacturing method immobilizes Protein A/G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • High capacity – “Plus” variety of Pierce Protein A/G Agarose has a very dense load of immobilized Protein A/G, providing a binding capacity greater than 50mg human IgG/mL resin

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Specifications

Product Type Agarose
Stationary Phase Protein A/G
Purification Target IgG Antibodies, Ig Antibodies
For Use With (Application) Antibody Purification
Content And Storage Protein A/G, Agarose Resin. Store in refrigerator (2–8°C).
Quantity 20 mL
Shipping Condition Wet Ice
Form Agarose Bead
I have noticed that there is a slightly higher amount of agarose resin in product Cat. No. 20424 than expected according to the manual. What is the reason for that?

To ensure the delivery of the correct amount of agarose resin, the products are slightly overfilled with resin. The slurry will be maintained at 50% so the resin and liquid solution volume will be equivalent.

What does it mean that the crosslinked 6% beaded agarose is supplied as a 50% slurry for products Pierce Protein A/G Plus Agarose (Cat. No. 20423 and 20424) and Pierce Protein A/G Agarose (Cat. No. 20421 and 20422)?

This means that the solution consists of 50% settled resin and 50% liquid solution. For example, Cat. No. 20424 contains 20 ml of settled resin and 20 ml of storage buffer, making it a 40 ml, 50% slurry.

For Research Use Only. Not for use in diagnostic procedures.