missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A Columns, 1 mL

Catalog No. PI20356
Change view
Click to view available options
Quantity:
5 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20356 5 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20356 Supplier Thermo Scientific™ Supplier No. LSG20356
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purify antibodies or perform immunoprecipitation with this standard-capacity, immobilized native Protein A on beaded agarose resin.

Perform a variety of antibody affinity purification methods with Thermo Scientific™ Pierce Protein A Agarose, a standard-capacity Protein A beaded agarose resin.

Pierce Protein A Agarose consists of purified native Protein A that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity purification procedure. Several package formats are offered: 5mL and 25mL bottles of resin slurry, 1mL pre-packed columns and a complete IgG purification kit.

Protein A is a bacteria-derived protein that binds with high affinity and specificity to the Fc portion of antibodies, especially those of the IgG class. Compared to its alternative (Protein G), Protein A provides the highest binding capacity for subclasses of IgG from rabbits, pigs, dogs and cats. Protein A also has higher binding capacity for human IgG, except for IgG3, which it binds weakly. Protein A also binds weakly to mouse IgG1 and is not recommended for most applications with that antibody isotype.

Highlights:

  • Native Protein A – immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum

  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods

  • Inert and stable – superior manufacturing method immobilizes Protein A by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield

  • Standard capacity – Pierce Protein A Agarose has a normal load of immobilized Protein A, providing a binding capacity of 12 to 19mg human IgG/mL resin (approx. 6 to 8mg mouse IgG/mL resin)

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Available Formats:

  • Resin Slurries: crosslinked 6% beaded agarose; 5mL and 25mL bottles
  • Spin Columns: 1mL columns
  • Purification Kit: complete kit containing five 1mL columns
  • Bulk and Custom: let us know your needs for bulk Pierce Protein A Agarose

Includes:

Kit contains Protein A Column (1mL each), Protein A IgG Binding Buffer, IgG Elution Buffer and desalting columns

Specifications

Product Type Column
Product Line Pierce
Column Type Agarose Resin, Affinity
Format 15 mL Conical Spin Column
Stationary Phase Protein A
Purification Target IgG Antibodies
Sufficient For 12 to 19 mg Human IgG/mL Column
Content And Storage Upon receipt store product at 4°C to 8°C. Product is shipped at ambient temperature.
Quantity 5 Columns

For Research Use Only. Not for use in diagnostic procedures.