Learn More
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-LC-Biotin
Description
Thermo Scientific Pierce Premium Grade Sulfo-NHS-LC-Biotin is our highest quality formulation of this popular amine-reactive biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount.
Features of Premium Grade Sulfo-NHS-LC-Biotin:
- Sulfo-NHS-LC-Biotin—popular amine-reactive biotinylation reagent for antibody labeling
- High quality—identity and purity confirmed by several tests, including quantitative NMR
- Product integrity—enhanced level of testing and characterization compared to standard grade
- Lot retention—ample supply of past lots retained to ensure future process testing
- Change management—Change Control Notification (CCN) service
- Consistent manufacture—batch-specific manufacturing documentation review
Compared to the standard grade product, Premium Grade Sulfo-NHS-LC-Biotin provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents.
Related Products
EZ-Link™ Sulfo-NHS-LC-Biotin (Cat. No. 21335)
Specifications
Specifications
| Product Type | Sulfo-NHS-LC-Biotin |
| Quantity | 100 mg |
| Content And Storage | Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature. |
| Product Line | Pierce |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, Sulfo-NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Label or Dye | Biotin |
| Spacer | Mid-length |
| Show More |
Frequently Asked Questions (FAQs)
Premium Grade Sulfo-NHS-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
For Research Use Only. Not for use in diagnostic procedures.