missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Phosphoprotein Phosphate Estimation Assay Kit

Catalog No. PI23270
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23270 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23270 Supplier Thermo Scientific™ Supplier No. LSG23270

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Classify and identify proteins as phosphorylated (serine and threonine) and obtain semi-quantitative assessment of the phosphorylation levels.

Thermo Scientific™ Phosphoprotein Phosphate Estimation Kit enables classification and identification of proteins as phosphorylated (serine and threonine), as well as semi-quantitative assessment of the phosphorylation level.

The Phosphoprotein Phosphate Estimation Assay Kit is designed to aid in characterization of the status and extent of phosphorylation of purified protein samples. The assay is based on the alkaline hydrolysis of phosphate from seryl and threonyl residues in phosphoproteins and quantification of the released phosphate with malachite green and ammonium molybdate. The assay is easily performed in 96-well microplates or test tubes and is completed in about one hour.

The assay can be used to identify whether a purified protein contains either phospho-serine (p-Ser) or phospho-threonine (p-Thr) as well as to estimate the level of this type of phosphorylation. For quantitation, the test protein sample is compared with specific concentrations of phosvitin, a phosphoprotein of known phosphorylation level. The alkaline hydrolysis step does not release phosphate from phospho-tyrosine (p-Tyr) residues in peptide linkage. Therefore, a negative result for an unknown purified protein preparation indicates that the protein is either (1) not a phosphoprotein or (2) is phosphorylated exclusively at tyrosine residues. In the latter case, Western blot analysis using an anti-phosphotyrosine antibody will be necessary to distinguish between these two possibilities.

Highlights:

  • Specific – measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only; does not measure phosphotyrosine (p-Tyr)

  • Convenient – test tube and 96-well microplate protocols included and require less than 90 minutes to perform
  • Semi-quantitative – calculate moles of phosphate (P) per mole of protein using the included phosvitin standard

  • Customizable – format is adaptable to development of specific assays for pure phosphoproteins when previously characterized standards are available

Includes:

Ammonium molybdate solution, malachite green solution, Tris-buffered saline pack and phopvitin positive control

Recommended for:

Identify proteins as containing phosphoserine or phosphothreonine phosphorylations; Estimate the amount of pS and pT phosphorylation; Develop specific quantitative assays for well-characterized phosphoproteins of interest

Specifications

Content And Storage Sufficient For: 500 tube assays or 1920 microplate assays
• Ammonium molybdate Solution, 25 mL
• Malachite Green Solution, 75 mL
• Phosvitin Positive Control, 1 mg
• TBS Pack (makes 500 mL), 1 pack

Upon receipt, store components at room temperature. Once dissolved, store Phosvitin solution at 4°C (or -20°C for long-term storage).
Assay Phosphoprotein Phosphate Estimation Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Phosphoprotein Phosphate Estimation
Specificity Not Target-Specific
Sufficient For 500 Tube Assays or 1920 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 100 mL
Do you have products to quantitate the degree of glycosylation or phosphorylation of my protein?

The Glycoprotein Carbohydrate Estimation Kit (Cat. No.23260) enables the amount of protein glycosylation to be measured as the percent of total purified protein mass.

The Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270) enables classification and identification of proteins as phosphorylated serine and threonine, as well as semi-quantitative assessment of the phosphorylation level.

Do you offer any protein assays that are protein specific?

We offer several “specialty” protein assays including: Protease assay kits (Cat. Nos. 23263 and 23266), Glycoprotein Carbohydrate Estimation Kit (Cat. No. 23260), Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270), Quantitative Peroxide assay kits (Cat. Nos. 23280 and 23285), and Easy-Titer Assay kits for IgG and IgM.

For Research Use Only. Not for use in diagnostic procedures.