missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Peroxidase Conjugate Stabilizer
Description
Thermo Scientific™ Pierce Peroxidase Conjugate Stabilizer is an antifreeze solution for storing aliquots and stocks of antibodies and other HRP-conjugates so they can be pipetted for use without thawing.
Highlights:
- Provides a buffered antifreeze environment for enzyme conjugated antibodies and proteins Maintains a liquid, low viscosity solution at -20°C
- No aliquoting of conjugate is necessary and sampling is convenient
- Contains preservatives to prevent anti-microbial growth during long-term storage
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. |
| Form | Liquid |
| Product Line | Pierce |
| Reagent Type | Stabilizing Reagent |
| Validated Application | ELISA, Immunohistochemistry, Western Blot |
| Quantity | 25 mL |
For Research Use Only. Not for use in diagnostic procedures.