Learn More
Thermo Scientific™ Pierce™ NeutrAvidin™ Coated Plates
Thermo Scientific™ Pierce NeutrAvidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Specifications
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Product Line NeutrAvidin Pierce
Material Polystyrene
Surface Treatment NeutrAvidin Protein
No. per Pack 5 Plates
View More Specs
Products 10
Catalog Number Mfr. No. Color No. of Wells Format Plate Blocking Price Quantity & Availability  
Catalog Number Mfr. No. Color No. of Wells Format Plate Blocking Price Quantity & Availability  
PI15217
View Documents Promotion Details
Thermo Scientific™
15217
Black 96 Standard Blocker™ BSA Buffer N/A
Only null left
PI15117
View Documents Promotion Details
Thermo Scientific™
LSG15117
Black 96 Standard SuperBlock™ Buffer N/A
Only null left
PI15402
View Documents Promotion Details
Thermo Scientific™
15402
Black 384 Standard SuperBlock™ Buffer N/A
Only null left
PI15128
View Documents Promotion Details
Thermo Scientific™
LSG15128
Clear 96 Strip Plate Blocker™ BSA Buffer N/A
Only null left
PI15123
View Documents Promotion Details
Thermo Scientific™
15123
Clear 96 Standard Blocker™ BSA Buffer N/A
Only null left
PI15129
View Documents Promotion Details
Thermo Scientific™
LSG15129
Clear 96 Standard SuperBlock™ Buffer N/A
Only null left
PI15400
View Documents Promotion Details
Thermo Scientific™
LSG15400
Clear 384 Standard SuperBlock™ Buffer N/A
Only null left
PI15216
View Documents Promotion Details
Thermo Scientific™
15216
White 96 Standard Blocker™ BSA Buffer N/A
Only null left
PI15116
View Documents Promotion Details
Thermo Scientific™
LSG15116
White 96 Standard SuperBlock™ Buffer N/A
Only null left
PI15401
View Documents Promotion Details
Thermo Scientific™
LSG15401
White 384 Standard SuperBlock™ Buffer N/A
Only null left
Description
Description
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. With a near-neutral pI (6.3), this results in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein lacks the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells.
Thermo Scientific NeutrAvidin Protein Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Features of NeutrAvidin Protein Coated Plates include:
  • White plates for chemiluminescent detection methods
  • Pre-blocked with Blocker BSA Buffer or SuperBlock Blocking Buffer
  • Easy and gentle immobilization of biotin-containing conjugates
  • Low nonspecific binding
  • No denaturing of the protein component of a conjugate upon binding
  • Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
  • Less expensive than streptavidin coated plates
  • Lowest nonspecific binding properties of all biotin-binding proteins
Specifications
Specifications
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Polystyrene
5 Plates
NeutrAvidin Pierce
NeutrAvidin Protein