missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NeutrAvidin™ Coated Plates

Catalog No. PI15129
Change view
Click to view available options
Color:
Black
Clear
White
No. of Wells:
384
96
Format:
Standard
Strip Plate
Plate Blocking:
Blocker™ BSA Buffer
SuperBlock™ Buffer
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells Format Plate Blocking
PI15129 Clear 96 Standard SuperBlock™ Buffer
PI15217 Black 96 Standard Blocker™ BSA Buffer
PI15117 Black 96 Standard SuperBlock™ Buffer
PI15402 Black 384 Standard SuperBlock™ Buffer
PI15128 Clear 96 Strip Plate Blocker™ BSA Buffer
PI15123 Clear 96 Standard Blocker™ BSA Buffer
PI15400 Clear 384 Standard SuperBlock™ Buffer
PI15216 White 96 Standard Blocker™ BSA Buffer
PI15116 White 96 Standard SuperBlock™ Buffer
PI15401 White 384 Standard SuperBlock™ Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. PI15129 Supplier Thermo Scientific™ Supplier No. LSG15129

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce NeutrAvidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.

Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. With a near-neutral pI (6.3), this results in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein lacks the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells.

Thermo Scientific NeutrAvidin Protein Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.

Features of NeutrAvidin Protein Coated Plates include:

  • White plates for chemiluminescent detection methods
  • Pre-blocked with Blocker BSA Buffer or SuperBlock Blocking Buffer
  • Easy and gentle immobilization of biotin-containing conjugates
  • Low nonspecific binding
  • No denaturing of the protein component of a conjugate upon binding
  • Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
  • Less expensive than streptavidin coated plates
  • Lowest nonspecific binding properties of all biotin-binding proteins

Specifications

Binding Property Standard Capacity
Format Standard
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Detection Method Colorimetric
Product Line NeutrAvidin Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
No. of Wells 96
Surface Treatment NeutrAvidin Protein
No. per Pack 5 Plates
Plate Blocking SuperBlock™ Buffer
Show More Show Less