missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BCIP Substrate Powder (5-bromo-4-chloro-3'-indolylphosphate p-toluidine)

Catalog No. PI34040
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34040 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI34040 Supplier Thermo Scientific™ Supplier No. 34040

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powders and ready-made nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolylphosphate solutions for chromogenic detection of alkaline phosphatase probes.

Thermo Scientific™ Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolylphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolylphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Highlights:

  • AP substrate – for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples

  • Chromogenic – no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph

  • Package options – choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

1-Step™ NBT/BCIP

  • Ideal for immunoblotting applications
  • Minimal assay-to-assay variability

1-Step NBT/BCIP Plus Suppressor

  • Ideal for staining tissues assayed with calf intestinal alkaline phosphatase without the interference caused by endogenous phosphatases
  • Contains 1mM levamisole to inhibit endogenous phosphatase activity

Specifications

Description Pierce BCIP Substrate
Product Type Substrate
Detection Method Colorimetric
Form Powder
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce
Quantity 1 g
Substrate Alkaline Phosphatase
Content And Storage Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
What alkaline phosphatase substrates do you offer?

For Research Use Only. Not for use in diagnostic procedures.