missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Modified Lowry Protein Assay Kit

Catalog No. PI23240
Change view
Click to view available options
Quantity:
530 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23240 530 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23240 Supplier Thermo Scientific™ Supplier No. LSG23240

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clear-to-purple (A750nm), 2-component, folin phenol- and copper-based reagent system to measure total protein concentration vs. protein standard.

Thermo Scientific™ Pierce Modified Lowry Protein Assay is a classical, well-characterized copper-based method for measuring protein concentration.

Additional Information and Online Ordering

The Modified Lowry Protein Assay Kit combines a stabilized formulation of the original Lowry Reagents and the essential Folin-Ciocalteu Phenol reagent in a complete kit for accurately determining protein concentration in a variety of samples types. Although newer protein assay methods provide greater speed and convenience, the Lowry method remains a popular, accurate, and useful option for many applications.

Highlights:

  • Popular method – widely cited in protein research literature

  • Colorimetric – measure with a standard spectrophotometer or plate reader (750nm)

  • Stable – uses a modified cupric sulfate-tartrate reagent that is stable at room temperature

  • Large range – exhibits good linearity in the range 1 to 1500μg/mL (tested with BSA protein)

  • Convenient – microplate and cuvette protocols provided with the instructions

Includes:

Modified Lowry Protein Assay Reagent, Folin-Ciocalteu Phenol Reagent (2N), and ampules of Albumin Standard (2mg/mL)

Specifications

Content And Storage Sufficient For: 480 tube assays or 2400 microplate assays

• Modified Lowry Protein Assay Reagent, 480 mL
• 2N Folin-Ciocalteu Phenol Reagent, 50 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL

Store at 4°C.
Assay Lowry Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 480 Tube Assays or 2400 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 530 mL

For Research Use Only. Not for use in diagnostic procedures.